siRNA Details
| Gene Symbol: Cpn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027904.3 | NP_082180.2 | 2 | 1344 | ||
| GenBank mRNA | AK005049.1 | - | 2 | 1346 | ||
| BC025836.1 | - | 2 | 1311 | |||
| BC081550.1 | - | 1 | 863 | |||
| BC138287.1 | - | 2 | 1431 | 8861306 | ||
| BC138288.1 | - | 2 | 1431 | 8861307 | ||

Cpn2
Genomic Map