siRNA Details
| Gene Symbol: Mtfr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027930.3 | NP_082206.2 | 6 | 676 | ||
| GenBank mRNA | AK011410.1 | - | 6 | 671 | ||
| AK016786.1 | - | 6 | 721 | |||
| BC019132.1 | - | 6 | 680 | 5036908 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mtfr2| Gene Symbol: Mtfr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027930.3 | NP_082206.2 | 6 | 676 | ||
| GenBank mRNA | AK011410.1 | - | 6 | 671 | ||
| AK016786.1 | - | 6 | 721 | |||
| BC019132.1 | - | 6 | 680 | 5036908 | ||