This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other 2310047M10RIK SIRNA Assays ›
Gene Symbol:
2310047M10Rik
Gene Name:
RIKEN cDNA 2310047M10 gene
Gene Aliases:
-
Chromosome Location:
Chr.11: 69059775 - 69061576 on Build GRCm38
UniGene ID:
Mm.340257
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s90310

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: 2310047M10Rik
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028005.3 NP_082281.2 1 962
GenBank mRNA AK009886.1 - 1 1005
AK151629.1 - 1 964
AK169875.1 - 1 943

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_017622, BC018880, AK000021, BC009261, BC070272, and others.
This siRNA also targets the following transcripts in Rat: NM_001017474, BC082026.

Gene Ontology Categories:

Function(s) Process(es)

biological_process
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-55586bc97b-jq955:80/100.66.72.86:80.
git-commit: d8c775737291b1e1a3c617895b4ca460ad723db2
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0