This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other 2310067B10RIK SIRNA Assays ›
Gene Symbol:
2310067B10Rik
Gene Name:
RIKEN cDNA 2310067B10 gene
Gene Aliases:
Kiaa0195, mKIAA0195
Chromosome Location:
Chr.11: 115765433 - 115799033 on Build GRCm38
UniGene ID:
Mm.23168
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s90337

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 5,354.00
Your Price
Online offer:
5354.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: 2310067B10Rik
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028014.3 NP_082290.2 15 1944
GenBank mRNA AK047622.1 - 1 78
AK155233.1 - 15 1819
BC007157.1 - 2 190
BC053088.1 - 15 1859 5709104

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-875b6bdcc-5twsw:80/100.66.79.81:80.
git-commit: 41fbaa8c9960493fb804edf63312caa25f4bdf16
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0