siRNA Details
| Gene Symbol: Cnn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028044.2 | NP_082320.1 | 7 | 961 | ||
| GenBank mRNA | AK005460.1 | - | 7 | 963 | ||
| AK051121.1 | - | 6 | 1179 | |||
| AK051628.1 | - | 7 | 979 | |||
| AK081672.1 | - | 7 | 876 | |||
| AK159555.1 | - | 7 | 1006 | |||
| AK159871.1 | - | 7 | 991 | |||
| AK166578.1 | - | 7 | 969 | |||
| AK167201.1 | - | 7 | 982 | |||
| AK167613.1 | - | 7 | 979 | |||
| AK169021.1 | - | 7 | 986 | |||
| AK169098.1 | - | 7 | 1006 | |||
| AK169410.1 | - | 7 | 936 | |||
| AK205558.1 | - | 1 | 66 | |||
| BC005788.1 | - | 3 | 288 | |||
| BC039643.1 | - | 3 | 356 | |||
| BC055711.1 | - | 7 | 962 | 6415164 | ||
| BC085268.1 | - | 7 | 739 | 6487559 | ||

Cnn3
Genomic Map