This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other 2010111I01RIK SIRNA Assays ›
Gene Symbol:
2010111I01Rik
Gene Name:
RIKEN cDNA 2010111I01 gene
Gene Aliases:
2300006M17Rik, AP-O, Aopep, mir-23b, mir-24-1, mir-27b
Chromosome Location:
Chr.13: 62964893 - 63326096 on Build GRCm38
UniGene ID:
Mm.188105
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s90546

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: 2010111I01Rik
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001289924.1 NP_001276853.1 2 939
NM_001289926.1 NP_001276855.1 2 939
GenBank mRNA AJ810420.1 - 1 671
AK044477.1 - 2 940
AK141931.1 - 1 905
BC042715.1 - 1 357
BC141279.1 - 1 790 9055891

Back To Top

More Information


Panther Classification:

Molecular Function -

protease protein modifying enzyme

Gene Ontology Categories:

Function(s) Process(es)

proteolysis
leukotriene biosynthetic process
aminopeptidase activity
peptidase activity
metallopeptidase activity
zinc ion binding
hydrolase activity
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-gb242:80/100.66.79.144:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline