This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other RUSC1 SIRNA Assays ›
Gene Symbol:
Rusc1
Gene Name:
RUN and SH3 domain containing 1
Gene Aliases:
2210403N08Rik, AA408288, NESCA
Chromosome Location:
Chr.3: 89083975 - 89093363 on Build GRCm38
UniGene ID:
Mm.27687
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s90832

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Rusc1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001083807.1 NP_001077276.1 4 1685
NM_001083808.1 NP_001077277.1 3 225
NM_028188.2 NP_082464.2 5 2096
GenBank mRNA AK032334.1 - 3 715
AK039664.1 - 5 2024
AK042689.1 - 4 1633
AK155528.1 - 1 246
AK170009.1 - 1 246
AK170012.1 - 3 240
AK170210.1 - 3 188
BC052081.1 - 3 429
BC056360.1 - 4 1623 6816267
BC057034.1 - 3 291 5687517
BY201665.1 - 3 226

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: XM_008761235, XM_008761232, XM_008761234, XM_008761233, XM_008761237, and others.

Gene Ontology Categories:

Function(s) Process(es)

protein polyubiquitination
actin binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-cj8gb:80/100.66.76.113:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline