This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other RUSC1 SIRNA Assays ›
Gene Symbol:
Rusc1
Gene Name:
RUN and SH3 domain containing 1
Gene Aliases:
2210403N08Rik, AA408288, NESCA
Chromosome Location:
Chr.3: 89083975 - 89093363 on Build GRCm38
UniGene ID:
Mm.27687
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s90833

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Rusc1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001083807.1 NP_001077276.1 10 2741
NM_001083808.1 NP_001077277.1 9 1281
NM_028188.2 NP_082464.2 11 3152
GenBank mRNA AK032334.1 - 9 1771
AK039664.1 - 11 3080
AK042689.1 - 10 2689
AK170012.1 - 9 1295
AK170210.1 - 9 1244
BC052081.1 - 8 1475
BC056360.1 - 10 2679 6816267
BC057034.1 - 9 1347 5687517

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

protein polyubiquitination
actin binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-dz54l:80/100.66.79.164:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline