This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other 2210016L21RIK SIRNA Assays ›
Gene Symbol:
2210016L21Rik
Gene Name:
RIKEN cDNA 2210016L21 gene
Gene Aliases:
-
Chromosome Location:
Chr.5: 114942163 - 114948544 on Build GRCm38
UniGene ID:
Mm.292672
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s90919

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: 2210016L21Rik
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028211.1 NP_082487.1 6 637
GenBank mRNA AK135000.1 - 655
BC031162.1 - 6 637 4982352

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001009630, BC088751.

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-cj8gb:80/100.66.76.113:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline