This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other TDRKH SIRNA Assays ›
Gene Symbol:
Tdrkh
Gene Name:
tudor and KH domain containing protein
Gene Aliases:
2700091C21Rik, Tdrd2
Chromosome Location:
Chr.3: 94413239 - 94432975 on Build GRCm38
UniGene ID:
Mm.331198
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s91166

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 400.00
Your Price
Online offer: 296.00
296.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Tdrkh
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028307.1 NP_082583.1 5, 6 642
GenBank mRNA AK012596.1 - 5, 6 683
BC031886.1 - 5, 6 536
BC049363.1 - 5, 6 642 6490411
BC057030.1 - 5, 6 658 6833743

Back To Top

More Information


Panther Classification:

Molecular Function -

scaffold/adaptor protein

Gene Ontology Categories:

Function(s) Process(es)

male meiosis
spermatogenesis
fertilization
cell differentiation
gene silencing by RNA
piRNA metabolic process
DNA methylation involved in gamete generation
nucleic acid binding
RNA binding
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-4gvqm:80/100.66.78.154:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline