This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other TONSL SIRNA Assays ›
Gene Symbol:
Tonsl
Gene Name:
tonsoku-like, DNA repair protein
Gene Aliases:
2810439M11Rik, Nfkbil2
Chromosome Location:
Chr.15: 76626237 - 76639929 on Build GRCm38
UniGene ID:
Mm.358749
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s91274

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Tonsl
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_183091.3 NP_898914.3 23 3731
GenBank mRNA AK013272.1 - 3 529
BC066068.1 - 23 3713 6851399

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

double-strand break repair via homologous recombination
replication fork processing
histone binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-f8k5r:80/100.66.76.16:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline