siRNA Details
| Gene Symbol: Cxx1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028375.3 | NP_082651.1 | 1 | 275 | ||
| GenBank mRNA | AK013603.1 | - | 1 | 166 | ||
| BC031701.1 | - | 1 | 163 | 3594379 | ||
| BY123946.1 | - | 1 | 275 | |||
| CK627323.1 | - | 1 | 192 | |||

Cxx1c
Genomic Map