siRNA Details
| Gene Symbol: Ttc17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183106.2 | NP_898929.2 | 5 | 612 | ||
| GenBank mRNA | CA328748.1 | - | 2 | 75 | ||
| CF536785.1 | - | 5 | 563 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ttc17| Gene Symbol: Ttc17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183106.2 | NP_898929.2 | 5 | 612 | ||
| GenBank mRNA | CA328748.1 | - | 2 | 75 | ||
| CF536785.1 | - | 5 | 563 | |||