siRNA Details
| Gene Symbol: Atp13a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164366.1 | NP_001157838.1 | 29 | 3587 | ||
| NM_029097.2 | NP_083373.2 | 29 | 3661 | |||
| GenBank mRNA | AK003623.1 | - | 13 | 1726 | ||
| AK156104.1 | - | 29 | 3640 | |||
| AK171305.1 | - | 29 | 3541 | |||
| AK214253.1 | - | 2 | 204 | |||
| AK216739.1 | - | 1 | 20 | |||
| BC023746.1 | - | 26 | 3963 | 5353766 | ||
| BC042661.1 | - | 29 | 3606 | 4192094 | ||

Atp13a2
Genomic Map