This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other 1700020N15RIK SIRNA Assays ›
Gene Symbol:
1700020N15Rik Gm6812
Gene Name:
RIKEN cDNA 1700020N15 gene
predicted gene 6812
Gene Aliases:
-
EG627927
Chromosome Location:
Chr.X: 69945281 - 69945980 on Build GRCm38
Chromosome Location:
Chr.X: 68892373 - 68893053 on Build GRCm38
UniGene ID:
Mm.233475
Mm.391264
Species:
Mouse
Species Specific ID (Flybase ID):
-
-
siRNA ID s93583

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: 1700020N15Rik
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029316.1 NP_083592.1 2 312
GenBank mRNA AK006180.1 - 2 294
BC049765.1 - 290
CA464409.1 - 2 314
Gene Symbol: Gm6812
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001098842.2 NP_001092312.1 2 293

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0