siRNA Details
| Gene Symbol: Akap13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029332.1 | NP_083608.1 | 7 | 2715 | ||
| GenBank mRNA | AK149507.1 | - | 1 | 80 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Akap13| Gene Symbol: Akap13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029332.1 | NP_083608.1 | 7 | 2715 | ||
| GenBank mRNA | AK149507.1 | - | 1 | 80 | ||