This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other SMURF1 SIRNA Assays ›
Gene Symbol:
Smurf1
Gene Name:
SMAD specific E3 ubiquitin protein ligase 1
Gene Aliases:
4930431E10Rik, mKIAA1625
Chromosome Location:
Chr.5: 144876495 - 144965830 on Build GRCm38
UniGene ID:
Mm.27735
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s93915

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 5,354.00
Your Price
Online offer:
5354.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Smurf1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001038627.1 NP_001033716.1 5 631
NM_029438.3 NP_083714.3 5 631
GenBank mRNA AK015264.1 - 1 30
AK154491.1 - 5 632
BC029097.1 - 5 659 3660965
CX568994.1 - 3 231

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001109598, XM_006248882, XM_008769002.

Panther Classification:

Molecular Function -

ubiquitin-protein ligase

Gene Ontology Categories:

Function(s) Process(es)

protein polyubiquitination
positive regulation of defense response to virus by host
protein export from nucleus
transforming growth factor beta receptor signaling pathway
protein ubiquitination
cell differentiation
negative regulation of ossification
BMP signaling pathway
negative regulation of transforming growth factor beta receptor signaling pathway
ubiquitin-dependent SMAD protein catabolic process
receptor catabolic process
protein localization to cell surface
protein ubiquitination involved in ubiquitin-dependent protein catabolic process
proteasome-mediated ubiquitin-dependent protein catabolic process
protein targeting to vacuole involved in autophagy
mitophagy in response to mitochondrial depolarization
positive regulation of dendrite extension
positive regulation of protein ubiquitination involved in ubiquitin-dependent protein catabolic process
ubiquitin-protein transferase activity
UDP-N-acetylmuramoylalanyl-D-glutamyl-2,6-diaminopimelate-D-alanyl-D-alanine ligase activity
ligase activity
ribosomal S6-glutamic acid ligase activity
coenzyme F420-0 gamma-glutamyl ligase activity
coenzyme F420-2 alpha-glutamyl ligase activity
ubiquitin protein ligase activity
I-SMAD binding
R-SMAD binding
protein-glycine ligase activity
protein-glycine ligase activity, initiating
protein-glycine ligase activity, elongating
tubulin-glycine ligase activity
protein-glutamic acid ligase activity
tubulin-glutamic acid ligase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0