siRNA Details
| Gene Symbol: Slc35d3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029529.3 | NP_083805.1 | 2 | 1366 | ||
| GenBank mRNA | AK018094.1 | - | 1 | 148 | ||
| AK042761.1 | - | 2 | 1118 | |||
| AK044973.1 | - | 2 | 1103 | |||
| AK047610.1 | - | 2 | 1367 | |||
| BC139194.1 | - | 2 | 1196 | 8862216 | ||

Slc35d3
Genomic Map