This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other 1700039E15RIK SIRNA Assays ›
Gene Symbol:
1700039E15Rik
Gene Name:
RIKEN cDNA 1700039E15 gene
Gene Aliases:
Gm19188
Chromosome Location:
Chr.7: 45277331 - 45288996 on Build GRCm38
UniGene ID:
Mm.425445
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s94590

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: 1700039E15Rik
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033176.1 NP_001028348.1 6 1210
GenBank mRNA AK133083.1 - 6 1210
BC100502.1 - 2 1012
BC147591.1 - 6 1307 9056581
BC147593.1 - 6 1307 9056583
BC172112.1 - 6 1248

Back To Top

More Information


Panther Classification:

Molecular Function -

primary active transporter

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-kbrl8:80/100.66.76.113:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline