This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other ARID2 SIRNA Assays ›
Gene Symbol:
Arid2
Gene Name:
AT rich interactive domain 2 (ARID, RFX-like)
Gene Aliases:
1700124K17Rik, 4432409D24Rik, zipzap/p200
Chromosome Location:
Chr.15: 96287522 - 96405463 on Build GRCm38
UniGene ID:
Mm.17166
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s94900

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 5,354.00
Your Price
Online offer:
5354.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Arid2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175251.4 NP_780460.3 7, 8 946
GenBank mRNA CK781204.1 - 3, 4 308

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: XM_345867.

Gene Ontology Categories:

Function(s) Process(es)

nucleosome disassembly
positive regulation of transcription from RNA polymerase II promoter
transcription factor binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-hssmq:80/100.66.76.147:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline