This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other TMEM116 SIRNA Assays ›
Gene Symbol:
Tmem116
Gene Name:
transmembrane protein 116
Gene Aliases:
4930406A18Rik, 4930513P12Rik, C030022K24Rik
Chromosome Location:
Chr.5: 121452709 - 121495421 on Build GRCm38
UniGene ID:
Mm.358781
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s95017

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Tmem116
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001161626.1 NP_001155098.1 8 630
NM_001161627.1 NP_001155099.1 9 735
NM_029912.2 NP_084188.1 8 668
GenBank mRNA AK015103.1 - 10 885
AK015791.1 - 693
AK021099.1 - 8 674
AK039773.1 - 1179
BC019151.1 - 8 625 5064772
CK127968.1 - 9 735
DV658154.1 - 6 469

Back To Top

More Information


Panther Classification:

Molecular Function -

G-protein coupled receptor

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-55586bc97b-stzfq:80/100.66.77.128:80.
git-commit: d8c775737291b1e1a3c617895b4ca460ad723db2
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0