siRNA Details
| Gene Symbol: Lsm6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191004.1 | NP_001177933.1 | 4 | 388 | ||
| NM_030145.3 | NP_084421.1 | 3 | 289 | |||
| GenBank mRNA | AK005329.1 | - | 1 | 230 | ||
| AK019126.1 | - | 4 | 348 | |||
| AK078392.1 | - | 4 | 338 | |||
| AK141884.1 | - | 4 | 335 | |||
| AK158236.1 | - | 3 | 278 | |||
| AK161686.1 | - | 4 | 342 | |||
| BC030427.1 | - | 3 | 236 | 5370660 | ||
| BC098230.1 | - | 3 | 239 | 3372313 | ||
| BC099484.1 | - | 3 | 294 | 30936207 | ||
| DV648841.1 | - | 4 | 388 | |||

Lsm6
Genomic Map