This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other STPG1 SIRNA Assays ›
Gene Symbol:
Stpg1
Gene Name:
sperm tail PG rich repeat containing 1
Gene Aliases:
4930403G18Rik, 4930555I21Rik, Mapo2
Chromosome Location:
Chr.4: 135494833 - 135537803 on Build GRCm38
UniGene ID:
Mm.374844
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s95732

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 5,354.00
Your Price
Online offer:
5354.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Stpg1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030189.3 NP_084465.1 7, 8 970
GenBank mRNA AK015061.1 - 8, 9 1164
AK019766.1 - 7, 8 971
BC145657.1 - 6, 7 757

Back To Top

More Information


Panther Classification:

Molecular Function -

structural protein

Gene Ontology Categories:

Function(s) Process(es)

apoptotic process
biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-wtj7v:80/100.66.79.197:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline