siRNA Details
| Gene Symbol: Wbscr27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024479.2 | NP_077799.2 | 5 | 607 | ||
| GenBank mRNA | AK081683.1 | - | 5 | 626 | ||
| AK132481.1 | - | 5 | 596 | |||
| AY138964.1 | - | 5 | 607 | |||
| AY354925.2 | - | 5 | 607 | |||
| AY354926.1 | - | 5 | 508 | |||
| BC002286.1 | - | 5 | 598 | 3589835 | ||

Wbscr27
Genomic Map