siRNA Details
| Gene Symbol: Car15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030558.2 | NP_085035.1 | 3 | 271 | ||
| GenBank mRNA | AF231122.1 | - | 3 | 223 | ||
| AK131988.1 | - | 3 | 299 | |||
| BC019975.1 | - | 3 | 271 | 4223351 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Car15| Gene Symbol: Car15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030558.2 | NP_085035.1 | 3 | 271 | ||
| GenBank mRNA | AF231122.1 | - | 3 | 223 | ||
| AK131988.1 | - | 3 | 299 | |||
| BC019975.1 | - | 3 | 271 | 4223351 | ||