siRNA Details
| Gene Symbol: Nedd4l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF277232.1 | - | 17, 18 | 1683 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nedd4l| Gene Symbol: Nedd4l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF277232.1 | - | 17, 18 | 1683 | ||