siRNA Details
| Gene Symbol: Strn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001172098.1 | NP_001165569.1 | 11 | 1644 | ||
| NM_052973.2 | NP_443205.1 | 13 | 1896 | |||
| GenBank mRNA | AF307777.1 | - | 13 | 1880 | ||
| AK140447.1 | - | 11 | 1632 | |||
| AK165908.1 | - | 11 | 1328 | |||
| BC138055.1 | - | 11 | 1595 | 8861074 | ||
| BC145127.1 | - | 11 | 1595 | 9053675 | ||
| EF685152.1 | - | 14 | 1797 | |||

Strn3
Genomic Map