This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other STK17B SIRNA Assays ›
Gene Symbol:
Stk17b
Gene Name:
serine/threonine kinase 17b (apoptosis-inducing)
Gene Aliases:
3110009A03Rik, AI120141, Drak2
Chromosome Location:
Chr.1: 53755512 - 53785250 on Build GRCm38
UniGene ID:
Mm.25559
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s97242

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 5,354.00
Your Price
Online offer:
5354.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Stk17b
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133810.3 NP_598571.2 3 412
GenBank mRNA AK035693.1 - 3 380
AK076932.1 - 3 379
AK083332.1 - 3 381
AK153108.1 - 3 356
AK165291.1 - 3 352
AY092028.1 - 3 365

Back To Top

More Information


Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Gene Ontology Categories:

Function(s) Process(es)

protein phosphorylation
apoptotic process
programmed cell death
phosphorylation
intracellular signal transduction
protein autophosphorylation
positive regulation of fibroblast apoptotic process
nucleotide binding
protein kinase activity
protein serine/threonine kinase activity
ATP binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-55658555d6-9rjdb:80/100.66.79.224:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline