siRNA Details
| Gene Symbol: Akna | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK029701.1 | - | 5 | 1024 | ||
| AK129468.1 | - | 2534 | ||||
| AK154663.1 | - | 11 | 2200 | |||
| AK158313.1 | - | 13 | 2896 | |||
| AK159547.1 | - | 3066 | ||||
| BC025835.1 | - | 400 | ||||
| BC036324.1 | - | 3 | 459 | |||
| BC092227.1 | - | 12 | 2845 | 30531788 | ||

Akna
Genomic Map