siRNA Details
| Gene Symbol: AW011738 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK036383.1 | - | 1 | 350 | ||
| AK044300.1 | - | 351 | ||||
| AK078660.1 | - | 1 | 351 | |||
| AK083998.1 | - | 384 | ||||
| AK085774.1 | - | 350 | ||||
| AK086145.1 | - | 1 | 370 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

AW011738| Gene Symbol: AW011738 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK036383.1 | - | 1 | 350 | ||
| AK044300.1 | - | 351 | ||||
| AK078660.1 | - | 1 | 351 | |||
| AK083998.1 | - | 384 | ||||
| AK085774.1 | - | 350 | ||||
| AK086145.1 | - | 1 | 370 | |||