This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other PRRT4 SIRNA Assays ›
Gene Symbol:
Prrt4
Gene Name:
proline-rich transmembrane protein 4
Gene Aliases:
AW455481, D330027H18Rik
Chromosome Location:
Chr.6: 29169230 - 29179584 on Build GRCm38
UniGene ID:
Mm.22818
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s97745

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 5,354.00
Your Price
Online offer:
5354.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Prrt4
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001101443.1 NP_001094913.1 4 1001
GenBank mRNA AK052319.1 - 4 1003
BC140954.1 - 4 997 9055566

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001109226, XM_008762817, XM_006236220.

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-55586bc97b-gm6kg:80/100.66.79.106:80.
git-commit: d8c775737291b1e1a3c617895b4ca460ad723db2
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0