This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other SPRED3 SIRNA Assays ›
Gene Symbol:
Spred3
Gene Name:
sprouty-related, EVH1 domain containing 3
Gene Aliases:
AW047143, D130060H24Rik, Spred-3
Chromosome Location:
Chr.7: 29157783 - 29169854 on Build GRCm38
UniGene ID:
Mm.377262
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s97833

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Spred3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_182927.3 NP_891557.3 2, 3 315
GenBank mRNA AB115762.1 - 1, 2 183
BC052757.1 - 2, 3 287
BC062119.1 - 2, 3 315 6821036

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: XM_008759146, XM_008759144, XM_008759145.

Panther Classification:

Molecular Function -

scaffold/adaptor protein

Gene Ontology Categories:

Function(s) Process(es)

inactivation of MAPK activity
multicellular organism development
regulation of signal transduction
stem cell factor receptor binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-7669c5b596-vc5lg:80/100.66.78.61:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0