siRNA Details
| Gene Symbol: Cenpi | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK043344.1 | - | 18, 19 | 2031 | ||
| AK145200.1 | - | 19, 20 | 2087 | |||
| AK161473.1 | - | 19, 20 | 2170 | |||
| BC024848.1 | - | 2, 3 | 683 | |||
| BC030328.1 | - | 19, 20 | 2087 | 5376020 | ||

Cenpi
Genomic Map