This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other E130012A19RIK SIRNA Assays ›
Gene Symbol:
E130012A19Rik
Gene Name:
RIKEN cDNA E130012A19 gene
Gene Aliases:
AA409164, AI413509, AW539173, E13, esPRC2p48
Chromosome Location:
Chr.11: 97627387 - 97629716 on Build GRCm38
UniGene ID:
Mm.24506
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s98096

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 5,354.00
Your Price
Online offer:
5354.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: E130012A19Rik
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175332.3 NP_780541.2 1 1780
GenBank mRNA AK087411.1 - 1 1767
BC006054.1 - 1 1219
BC055770.1 - 1 1780 6403695

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

neuron fate commitment
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-584499df9d-2m6b5:80/100.66.73.235:80.
git-commit: 256cec658627a60c8e1196038bf5905a13cd45ba
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.33.0-2025.07.30-1.0