siRNA Details
| Gene Symbol: Ndufb11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019435.4 | NP_062308.2 | 3 | 710 | ||
| GenBank mRNA | AK028121.1 | - | 3 | 431 | ||
| BC027265.1 | - | 3 | 703 | 3589704 | ||
| BC096047.1 | - | 3 | 406 | |||
| DV654320.1 | - | 3 | 598 | |||
| Y08702.1 | - | 3 | 403 | |||

Ndufb11
Genomic Map