siRNA Details
| Gene Symbol: G6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033221.3 | NP_001028393.1 | 2 | 323 | ||
| NM_001191012.1 | NP_001177941.1 | 2 | 323 | |||
| GenBank mRNA | AK137516.1 | - | 2 | 324 | ||
| GU269270.1 | - | 2 | 309 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

G6b| Gene Symbol: G6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033221.3 | NP_001028393.1 | 2 | 323 | ||
| NM_001191012.1 | NP_001177941.1 | 2 | 323 | |||
| GenBank mRNA | AK137516.1 | - | 2 | 324 | ||
| GU269270.1 | - | 2 | 309 | |||