This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other OLFR153 SIRNA Assays ›
Gene Symbol:
Olfr153 Olfr1131
Gene Name:
olfactory receptor 153
olfactory receptor 1131
Gene Aliases:
GA_x6K02T2Q125-49033418-49034341, MOR177-5, Olfr4-2, V5
MOR177-4
Chromosome Location:
Chr.2: 87532035 - 87532958 on Build GRCm38
Chromosome Location:
Chr.2: 87628465 - 87629394 on Build GRCm38
UniGene ID:
Mm.424751
Mm.377615
Species:
Mouse
Species Specific ID (Flybase ID):
-
-
siRNA ID s99813

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 400.00
Your Price
Online offer: 296.00
296.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Olfr153
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_206823.1 NP_996559.1 1 838
GenBank mRNA BC147769.1 - 1 1077 9056708
BC147777.1 - 1 1077 9056711
Gene Symbol: Olfr1131
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146658.2 NP_666869.2 1 838

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001000664, NM_001000327.

Panther Classification:

Molecular Function -

G-protein coupled receptor

Gene Ontology Categories:

Function(s) Process(es)

G-protein coupled receptor signaling pathway
sensory perception of smell
detection of chemical stimulus involved in sensory perception of smell
G-protein coupled receptor activity
olfactory receptor activity
odorant binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-jx6zp:80/100.66.77.18:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline