Learn More
Thermo Scientific™ 0.1 mL Individual UTW Tubes
Reduce incubation times during PCR with these ultra-thin wall individual PCR tubes with attached flat caps.
Specifications
Closure Color Clear
Description With attached optical flat caps
Type Tubes
View More Specs
Products 1
Catalog Number Mfr. No. Color Price Quantity & Availability  
Catalog Number Mfr. No. Color Price Quantity & Availability  
TUC0011
View Documents
Thermo Scientific™
TUC0011
White This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description
Description
UTW vessels have walls that are 0.15 mm thin where it matters the most - at the contact point between the thermal cycler block and the reactants. Thinner walls offer less thermal resistance for transferring heat from the block to the sample within the tube. This results in the ability to perform very rapid protocols by reducing incubation times during PCR.
Highlights
  • Caps form a secure seal, yet are easy to open and close
  • Compatible with thermal cyclers accepting low-profile 0.2 mL tubes
  • Ultra Thin Wall (UTW) low profile for fast PCR applications, compatible with the 24-well Piko Thermal Cycler and PikoReal Real-Time PCR System
Specifications
Specifications
Clear
With attached optical flat caps
Tubes
SDS
Certified free from human DNA, RNases, and DNases.