Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

0.8 Percent Platinum - 0.3 Percent Molybdenum on Carbon Powder, Reduced, Water Wet, 23 microns, BASF Catalyst, Spectrum™

Click to view available options
Quantity:
5 g
25 g
100 g
7440-06-4
Pt
195.08
BASFCYQUMIYNBI-UHFFFAOYSA-N
platinum
[Pt]
7440-06-4
Pt
195.08
BASFCYQUMIYNBI-UHFFFAOYSA-N
platinum
[Pt]
Additional Information Support: Carbon powder
Molybdenum (Mo) 0.008
Particle Size 23 microns
Platinum (Pt) 0.008
Quantity 5 g

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.