Learn More
CERILLIANT™ Chlorpheniramine-D6 maleate Solution, 100 μg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer:  MilliporeSigma Supelco C0861ML
Catalog No. 01-001-064
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
This internal standard is suitable for quantitation of chlorphenamine levels in urine, serum, or plasma by LC/MS or GC/MS for a variety of applications from clinical toxicology and pharmaceutical research to isotope dilution methods. Chlorphenamine is a first-generation antihistamine prescribed for the prevention of allergic conditions such as rhinitis.
Specifications
Specifications
67-56-1, 1219806-45-7
MFCD09840267
100 μg/mL in methanol (as free base)
Ampule
1 mL
C20D6H17ClN2O4
UN 1230 - class 3 - PG 2 - Methanol, solution
396.90
-20°C
9.7°C (49.5°F) - closed cup
Safety and Handling
Safety and Handling
H331-H370
H311
EINECSNumber : 200-659-6
SDS
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
CERILLIANT is a registered trademark of Cerilliant Corporation