Learn More
CERILLIANT™ N-Desmethylclobazam-8-chloro isomer-13C6 Solution, 100 μg/mL in Acetonitrile, Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer:  MilliporeSigma Supelco D0941ML
Catalog No. 01-001-213
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
An internal standard for quantification of N-desmethylclobazam levels in urine or other matrices by LC/MS or GC/MS for clinical toxicology, urine drug testing, or forensic analysis applications. Also known as norclobazam, this pharmacologically active biomarker is a major metabolite of clobazam. Clobazam is a benzodiazepine marketed under the brand names Frisium, Urbanol, and Onfi™ for use as an anxiolytic and anticonvulsant.
Specifications
Specifications
75-05-8
UN 1648 3 / PGII
292.67
-20°C
2°C (35.6°F)
13C6C9H11ClN2O2
100 μg/mL in acetonitrile
Ampule
1 mL
Safety and Handling
Safety and Handling
H332-H319
H312
EINECSNumber : 200-835-2
SDS
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
CERILLIANT is a registered trademark of Cerilliant Corporation