Learn More
CERILLIANT™ (+/-)-4-Fluoroamphetamine-D5 (side chain) Hydrochloride Solution, 100 μg/mL, Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer:  MilliporeSigma Supelco F0221ML
Catalog No. 01-001-357
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
A fluoro analog of amphetamine, 4-Fluoroamphetamine is sold in the illicit market as a club/designer drug under street names including R2D2, Flux and Flits. The drug produces stimulant and possibly empathogenic or psychoactive effects. This internal standard is suitable for numerous applications in GC/MS or LC/MS testing from forensic analysis and clinical toxicology to urine drug monitoring and isotope dilution methods.
Specifications
Specifications
67-56-1
UN 1230 - class 3 - PG 2 - Methanol, solution
194.69
-20°C
9.7°C (49.5°F)
C9H7D5FN · HCl
100 μg/mL in methanol (as free base)
Ampule
1 mL
Safety and Handling
Safety and Handling
H331-H370
H311
EINECSNumber : 200-659-6
SDS
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
CERILLIANT is a registered trademark of Cerilliant Corporation