Learn More
CERILLIANT™ Morphine N-oxide Solution, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer:  MilliporeSigma Supelco IMPM005011ML
Catalog No. 01-001-515
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
A Certified Snap-N-Spike™ Solution applicable for use in morphine impurity identification by LC-MS/MS or GC/MS. Morphine n-oxide is a minor urinary metabolite of morphine as well as a potential degradation product of morphine. Morphine is a potent opiate analgesic drug used to relieve both acute and chronic severe pain.
Specifications
Specifications
67-56-1, 639-46-3
UN 1230 - class 3 - PG 2 - Methanol, solution
301.34
-20°C
9.7°C (49.5°F) - closed cup
C17H19NO4
100 μg/mL in methanol
Ampule
1 mL
Safety and Handling
Safety and Handling
H331-H370
H311
EINECSNumber : 200-659-6
SDS
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
CERILLIANT is a registered trademark of Cerilliant Corporation