Learn More
CERILLIANT™ Nalbuphine-D3 Solution, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer: MilliporeSigma Supelco N0561ML
Description
An internal standard suitable for quantitation of nalbuphine levels by GC/MS or LC/MS for urine drug testing, clinical toxicology or forensic analysis. Nalbuphine is a semisynthetic opioid analgesic sold under the trade name Nubain™. This painkiller is prescribed for the relief of moderate to severe pain.Specifications
67-56-1 | |
UN 1230 - class 3 - PG 2 - Methanol | |
360.46 | |
-20°C | |
9.7°C (49.5°F) - closed cup |
C21H24D3NO4 | |
100 μg/mL in methanol | |
Ampule | |
1 mL |
Safety and Handling
H331-H370-H412
H311
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation