Learn More
CERILLIANT™ (+)-Norpropoxyphene maleate Solution, 1.0 mg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer:  MilliporeSigma Supelco N9131ML
Catalog No. 01-001-792
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
Norpropoxyphene is a major urinary metabolite of the opioid analgesic drug propoxyphene. Norpropoxyphene's toxicity makes the parent drug more likely to cause death from overdose compared to similar mild opioid analgesics. This analytical reference standard is suitable for use as a starting material in the preparation of calibrators or controls for LC/MS or GC/MS testing methods in clinical toxicology, urine drug testing, forensic analysis or pain prescription monitoring.
Specifications
Specifications
67-56-1, 159208-83-0
UN 1230 - class 3 - PG 2 - Methanol, solution
441.52
2 to 8°C
9.7°C (49.5°F) - closed cup
C25H31NO6
1.0 mg/mL in methanol (as free base)
Ampule
1 mL
Safety and Handling
Safety and Handling
H331-H370
H311
EINECSNumber : 200-659-6
SDS
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
CERILLIANT is a registered trademark of Cerilliant Corporation