Learn More
CERILLIANT™ Pentedrone Hydrochloride, 1.0 mg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer:  MilliporeSigma Supelco P0871ML
Catalog No. 01-001-861
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
Pentedrone is a bath salt cathinone and designer drug with stimulant effects. Since 2010, this illicit drug has been found in a number of bath salt mixes sold as legal highs at head shops and on the internet.
Specifications
Specifications
67-56-1, 879669-95-1
MFCD27953553
1.0 mg/mL in methanol (as free base)
227.73
-20°C
9.7°C (49.5°F) - closed cup
C12H17NO · HCl
UN 1230 - class 3 - PG 2 - Methanol, solution
Pentedrone hydrochloride solution
Ampule
1 mL
Safety and Handling
Safety and Handling
H331-H370
H311
SDS
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
CERILLIANT is a registered trademark of Cerilliant Corporation