Learn More
CERILLIANT™ Tizanidine-D4 Hydrochloride Solution, 100 μg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer:  MilliporeSigma Supelco T1201ML
Catalog No. 01-002-090
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
This stable-labeled internal standard is suitable for quantitation of tizanidine levels in GC/MS or LC/MS methods for clinical toxicology, urine drug testing, pain prescription monitoring, forensic analysis or isotope dilution applications. Tizanidine, sold under the tradename Zanaflex™, is a skeletal muscle relaxant used for treatment of spasticity associated with multiple sclerosis, and for painful muscle spasms.
Specifications
Specifications
67-56-1, 1188263-51-5
MFCD09264522
100 μg/mL in methanol (as free base)
Ampule
1 mL
C9H4D4ClN5S · HCl
UN 1230 - class 3 - PG 2 - Methanol, solution
294.20
-20°C
9.7°C (49.5°F) - closed cup
Safety and Handling
Safety and Handling
H331-H370
H311
SDS
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
CERILLIANT is a registered trademark of Cerilliant Corporation