Learn More
Heidolph™ Reactor-Ready™ Lid
Click to view available options
5 Neck
Description
Specifications
Includes | Rodaviss Sealing Caps |
Type | Reactor-Ready Lid |
Material | Borosilicate Glass (3.3) |
Certifications/Compliance | Glass meets international standards DIN/ISO 3585. PTFE Material complies with FDA21 CFR 177.1550, certified in accordance with and meets USP 27, NF22, 2004 for USP class VI plastics |
For Use With (Equipment) | Reactor-Ready™ Lab Reactors |
Joint Type | Joints: B19 (1), B24 (2), B29 (1), B34 (1) |
Autoclavable | Autoclavable |
Positions | 5 Neck |