Learn More
Eppendorf™ ep Dualfilter T.I.P.S.™ LoRetention Tips
Eppendorf™ ep Dualfilter T.I.P.S.™ LoRetention Tips have an ultrahomogeneous, and ultrahydrophobic surface for maximum retention and reproducibility
Supplier:  Eppendorf™ 0030078632
 View more versions of this product
Catalog No. 05-413-959
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
View alternate product
Description
Description
  • Designed for maximum recovery and reproducibility, ep Dualfilter T.I.P.S. LoRetention tips have an ultrahomogeneous and ultrahydrophobic surface
  • Significantly reduced foam formation during pipetting
  • No sample loss, reduced costs, and improved results
  • Ultrahomogenous surface for maximized reproducibility
  • Ultrahydrophobic surface for minimal sample loss
  • Extreme resistance to chemicals
  • Available in PCR clean and Eppendorf Quality
  • No coating
  • Modern slim rack design with reclosable lid
  • Improved sustainability with 20% less polypropylene consumed per rack
TRUSTED_SUSTAINABILITY
Specifications
Specifications
0.1 to 10 μL
Sterile
LoRetention
Lot-Specific Certificates Available
Rack
Dark Gray
ep DualFilter T.I.P.S
Filtered
Clear
34 mm
PCR Clean
Consumables
Air-Cushion
Polypropylene
Product Suggestions
Product Suggestions