Alfa Aesar™ 1,1,1,2,2,3,3-Heptafluoro-7,7-dimethyl-4,6-octanedione, 97%

Manufacturer: Alfa Aesar™ A1207722

Quantity 100g

 View more versions of this product

Catalog No. AAA1207722

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 17587-22-3
    Molecular Formula C10H11F7O2
    Formula Weight 296.19
    MDL Number MFCD00000435
    UN Number UN1224
    Beilstein 2056991
    Synonym 7,7-Dimethyl-1,1,1,2,2,3,3-heptafluoro-4,6-octanedione; Fod-H
    Chemical Name or Material 1,1,1,2,2,3,3-Heptafluoro-7,7-dimethyl-4,6-octanedione

    Specifications

    Formula Weight 296.19
    TSCA TSCA
    Molecular Formula C10H11F7O2
    CAS 17587-22-3
    Recommended Storage 2°C to 8°C
    Quantity 100g
    Health Hazard 3 GHS P Statement
    P280g-P305+P351+P338
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: KETONES, LIQUID, N.O.S.
    Flash Point 53°C (127°F)
    MDL Number MFCD00000435
    Beilstein 2056991
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H226-H315-H319-H335
    Flammable liquid and vapor.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    UN Number UN1224
    Synonym 7,7-Dimethyl-1,1,1,2,2,3,3-heptafluoro-4,6-octanedione; Fod-H
    Boiling Point 56° to 57°C (10mmHg)
    Chemical Name or Material 1,1,1,2,2,3,3-Heptafluoro-7,7-dimethyl-4,6-octanedione
    Assay 97%

    GHS H Statement
    H226-H315-H319-H335
    Flammable liquid and vapor.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    2°C to 8°C

    GHS P Statement
    P280g-P305+P351+P338
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit